N-Methyl-bis-trifluoroacetamide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:685-27-8
:
Formula:C₅H₃F₆NO₂
MDL Number:MFCD00000412
:223.07 g/mol
Boiling Pt:123…124 °C
Density:1.547 g/cm³ (25 °C)
Flash Pt:42 °C
Storage Temperature:Ambient
EINECS:211-680-5
UN:2920
ADR:8+3,II
:GHS02
:GHS05