Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-4-chloro-5-fluoroaniline
2-Bromo-4-chloro-5-fluoroaniline 98%
2-Bromo-4-chloro-5-fluoroaniline 98%
Irritant
2-Bromo-4-chloro-5-fluoroaniline 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Bromo-4-chloro-5-fluoroaniline 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
120694-11-3
:
Formula:
C₆H₄BrClFN
MDL Number:
MFCD12026231
:
224.458 g/mol
:
GHS07
Storage Temperature:
Ambient