Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2'-Methoxy-5'-(trifluoromethyl)acetophenone
2'-Methoxy-5'-(trifluoromethyl)acetophenone 98%
2'-Methoxy-5'-(trifluoromethyl)acetophenone 98%
2'-Methoxy-5'-(trifluoromethyl)acetophenone 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2'-Methoxy-5'-(trifluoromethyl)acetophenone 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
503464-99-1
:
Formula:
C₁₀H₉F₃O₂
MDL Number:
MFCD09832300
:
218.17 g/mol
Melting Pt:
45…47 °C
:
GHS07
Storage Temperature:
Ambient