Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Ethoxy-3-(trifluoromethyl)benzoyl chloride
4-Ethoxy-3-(trifluoromethyl)benzoyl chloride 97%
4-Ethoxy-3-(trifluoromethyl)benzoyl chloride 97%
4-Ethoxy-3-(trifluoromethyl)benzoyl chloride 97%
Supplier:
Apollo Scientific
Certificates
Expand image
4-Ethoxy-3-(trifluoromethyl)benzoyl chloride 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
647010-69-3
:
Formula:
C₁₀H₈ClF₃O₂
MDL Number:
MFCD13196580
:
252.62 g/mol
Storage Temperature:
Ambient