Order Entry
Germany
ContactUsLinkComponent
(2,6-Difluoro-3-methoxyphenyl)acetic acid 97%
(2,6-Difluoro-3-methoxyphenyl)acetic acid 97%
Catalog # APOSPC302380-1G
Supplier:  Apollo Scientific
CAS Number:  
(2,6-Difluoro-3-methoxyphenyl)acetic acid 97%
Catalog # APOSPC302380-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :186.16 g/mol
Melting Pt:124…126 °C
Storage Temperature:Ambient
CAS Number:886498-65-3
Formula:(C₆H₂F₂OCH₃)CH₂COOH
MDL Number:MFCD04115908