Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4'-Fluoro-3'-methylacetophenone
4'-Fluoro-3'-methylacetophenone 95%
4'-Fluoro-3'-methylacetophenone 95%
4'-Fluoro-3'-methylacetophenone 95%
Supplier:
Apollo Scientific
Certificates
Expand image
4'-Fluoro-3'-methylacetophenone 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
369-32-4
:
Formula:
C₉H₉FO
MDL Number:
MFCD04972879
:
152.17 g/mol
:
GHS07
Storage Temperature:
Ambient