
Ethyl-2-hydroxy-4-fluorobenzoate
Catalog # APOSPC300809-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:184.17 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1737-21-9
Formula:C₉H₉FO₃
MDL Number:MFCD06205199