2',4'-Difluoropropiophenone 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:85068-30-0
:
Formula:C₉H₈F₂O
MDL Number:MFCD00015507
:170.16 g/mol
Boiling Pt:51…52 °C/2mmHg
Density:1.193 g/cm³ (25 °C)
Flash Pt:76 °C
EINECS:285-291-4
:GHS07
Storage Temperature:Ambient