Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
trans-2,4-Difluorocinnamic acid
trans-2,4-Difluorocinnamic acid 98%
trans-2,4-Difluorocinnamic acid 98%
trans-2,4-Difluorocinnamic acid 98%
Supplier:
Apollo Scientific
Certificates
Expand image
trans-2,4-Difluorocinnamic acid 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
94977-52-3
Formula:
C₉H₆F₂O₂
MDL Number:
MFCD00010317
Melting Pt:
215…216 °C
:
184.14 g/mol
:
GHS07
Storage Temperature:
Ambient