Order Entry
Germany
Orders LinkContactUsLinkComponent
3,5-Difluorobenzophenone 97%

3,5-Difluorobenzophenone 97%

Irritant
Supplier:   Apollo Scientific
3,5-Difluorobenzophenone 97%
Supplier:  Apollo Scientific
Certificates
3,5-Difluorobenzophenone 97%

3 Options Available Below

Product Details & Documents
Synonyms:  (3,5-Difluorophenyl)(phenyl)methanone
  • CAS Number:179113-89-4
  • Formula:C₁₃H₈F₂O
  • MDL Number:MFCD00061140
  • Molecular Weight:218.2 g/mol
  • Boiling Pt:117 °C/5mmHg
  • Melting Pt:57…59 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,80 €
    21,80 €
    1 * 5 g
       21,80 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 69,50 €
    69,50 €
    1 * 25 g
       69,50 €

  • Bottle
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 116,00 €
    116,00 €
    1 * 50 g
       116,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.