2,4'-Difluorobenzophenone 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:342-25-6
:
Formula:C₁₃H₈F₂O
MDL Number:MFCD00000319
:218.2 g/mol
Boiling Pt:176…178 °C/16mmHg
Melting Pt:22…24 °C
Density:1.244 g/cm³ (25 °C)
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:206-441-7
UN:3077
ADR:9+EHS,III
:GHS07
:GHS09