2-Aminomethyl-5-fluorobenzeneboronic acid hydrochloride
Catalog # APOSPC2668-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:205.42 g/mol
Melting Pt:152…158 °C
Storage Temperature:Ambient
CAS Number:850568-03-5
Formula:C₇H₉BFNO₂·HCl
MDL Number:MFCD04115666