2-Bromo-3,5-bis(trifluoromethyl)aniline 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:174824-16-9
:
Formula:C₈H₄BrF₆N
MDL Number:MFCD00134419
:308.02 g/mol
Melting Pt:33…37 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06