
1,5-Dibromo-1,1,3,3,5,5-hexafluoropentane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:371-83-5
:
Formula:C₅H₄Br₂F₆
MDL Number:MFCD00236657
:337.89 g/mol
Boiling Pt:54.2 °C/14mmHg
Density:2.003 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient