Order Entry
Germany
ContactUsLinkComponent
 
Irritant
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid pinacol ester
Catalog # APOSPC200435-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid pinacol ester
Catalog # APOSPC200435-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :280.0997 g/mol
Melting Pt:66…71 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:603122-79-8
Formula:C₁₄H₁₈BFO₄
MDL Number:MFCD11855962