Ethyl-4-bromo-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1240278-21-0
:
Formula:C₇H₆BrF₃N₂O₂
MDL Number:MFCD13193102
Melting Pt:131…133 °C
:287.036 g/mol
Storage Temperature:Ambient