Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Chloro-4-fluoroaniline
3-Chloro-4-fluoroaniline 98%
3-Chloro-4-fluoroaniline 98%
:
Apollo Scientific
3-Chloro-4-fluoroaniline 98%
:
Apollo Scientific
:
367-21-5
:
:
C₆H₅ClFN
:
MFCD00007767
:
145.56 g/mol
:
227…228 °C
:
42…47 °C
:
1.42 g/cm³ (25 °C)
:
149 °C
:
Ambient
:
206-682-8
:
2811
:
6.1,III
:
GHS06