Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Chloro-2,6-dibromo-4-fluoroaniline
3-Chloro-2,6-dibromo-4-fluoroaniline 97%
3-Chloro-2,6-dibromo-4-fluoroaniline 97%
Irritant
3-Chloro-2,6-dibromo-4-fluoroaniline 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-Chloro-2,6-dibromo-4-fluoroaniline 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
175135-09-8
:
Formula:
C₆H₃Br₂ClFN
MDL Number:
MFCD00042199
:
303.36 g/mol
Melting Pt:
97…99 °C
:
GHS07
Storage Temperature:
Ambient