

4-Bromo-1,1,2-trifluoro-1-butene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:10493-44-4
:
Formula:C₄H₄BrF₃
MDL Number:MFCD00039274
Boiling Pt:95…98 °C
Density:1.639 g/cm³ (25 °C)
Flash Pt:17 °C
UN:1993
ADR:3,II
:188.97 g/mol
:GHS02
:GHS07
Storage Temperature:Ambient