
Bromopentafluorobenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:344-04-7
:
Formula:C₆BrF₅
MDL Number:MFCD00000287
:246.96 g/mol
Boiling Pt:137 °C
Melting Pt:–31 °C
Density:1.98 g/cm³ (25 °C)
Flash Pt:87 °C
EINECS:206-449-0
:GHS07
Storage Temperature:Ambient