Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Fluoro-N-methylaniline
2-Fluoro-N-methylaniline
2-Fluoro-N-methylaniline
Irritant
2-Fluoro-N-methylaniline
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Fluoro-N-methylaniline
Supplier:
Apollo Scientific
About this item
CAS Number:
1978-38-7
:
Formula:
C₇H₈FN
MDL Number:
MFCD00028030
Density:
1.1 g/cm³ (25 °C)
:
125.15 g/mol
:
GHS07
Storage Temperature:
Ambient