Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Fluoro-o-toluidine
6-Fluoro-o-toluidine 97%
6-Fluoro-o-toluidine 97%
:
Apollo Scientific
6-Fluoro-o-toluidine 97%
:
Apollo Scientific
:
443-89-0
:
:
C₇H₈FN
:
MFCD06658252
:
125.1435 g/mol
:
88…89 °C/13mmHg
:
1.082 g/cm³ (25 °C)
:
77 °C
:
Refrigerator
:
2811
:
6.1,III
:
GHS05
:
GHS07