Order Entry
Germany
ContactUsLinkComponent
2-Bromo-α,α,α-trifluorotoluene 99%
2-Bromo-α,α,α-trifluorotoluene 99%
Catalog # APOSPC1330-100G
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-α,α,α-trifluorotoluene 99%
Catalog # APOSPC1330-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :225.01 g/mol
Boiling Pt:167…168 °C
Density:1.652 g/cm³ (25 °C)
Flash Pt:52 °C
Storage Temperature:Ambient
EINECS:206-879-9
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:392-83-6
Formula:C₇H₄BrF₃
MDL Number:MFCD00000373