Ethyl-1-[[3-chloro-5-(trifluoromethyl)-2-pyridinyl](methyl)amino]-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxyl
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:338965-01-8
:
Formula:C₁₆H₁₇ClF₃N₃O₃
MDL Number:MFCD01568671
:391.78 g/mol
Melting Pt:75…77 °C
Storage Temperature:Ambient