
4,4,4-Trifluoro-1-(4-methoxyphenyl)butane-1,3-dione 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:15191-68-1
:
Formula:C₁₁H₉F₃O₃
MDL Number:MFCD03238723
:246.19 g/mol
Melting Pt:57…58 °C
:GHS07
Storage Temperature:Ambient