0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
4-Allyl-2,3,5,6-tetrafluorobenzoic acid
4-Allyl-2,3,5,6-tetrafluorobenzoic acid 97%
4-Allyl-2,3,5,6-tetrafluorobenzoic acid 97%
APOSPC1006T-250MG
:
Apollo Scientific
:
4-Allyl-2,3,5,6-tetrafluorobenzoic acid 97%
APOSPC1006T-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
234.15 g/mol
:
89…90 °C
:
GHS07
:
Ambient
:
79538-02-6
:
C₁₀H₆F₄O₂
:
MFCD00082610