
2,3,5,6-Tetrafluoro-p-toluic acid
Catalog # APOSPC10050-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:208.11 g/mol
Melting Pt:173…175 °C
:GHS07
Storage Temperature:Ambient
CAS Number:652-32-4
Formula:C₈H₄F₄O₂
MDL Number:MFCD00012302