Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Hydroxy-2-(trifluoromethyl)pyridine
5-Hydroxy-2-(trifluoromethyl)pyridine 97%
5-Hydroxy-2-(trifluoromethyl)pyridine 97%
Irritant
5-Hydroxy-2-(trifluoromethyl)pyridine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-Hydroxy-2-(trifluoromethyl)pyridine 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
216766-12-0
:
Formula:
C₆H₄F₃NO
MDL Number:
MFCD06801325
:
163.1 g/mol
Melting Pt:
173…176 °C (dec.)
:
GHS07
Storage Temperature:
Ambient