Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3,4,5-Tetrafluorobenzyl alcohol
2,3,4,5-Tetrafluorobenzyl alcohol, Technical Grade
2,3,4,5-Tetrafluorobenzyl alcohol, Technical Grade
2,3,4,5-Tetrafluorobenzyl alcohol, Technical Grade
Supplier:
Apollo Scientific
Certificates
Expand image
2,3,4,5-Tetrafluorobenzyl alcohol, Technical Grade
Supplier:
Apollo Scientific
About this item
CAS Number:
53072-18-7
:
Formula:
C₇H₄F₄O
MDL Number:
MFCD01631452
:
180.1 g/mol
Storage Temperature:
Ambient