4-Methylthio-3-nitrobenzotrifluoride
Catalog # APOSPC0463-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Melting Pt:94…96 °C
Storage Temperature:Ambient
CAS Number:71236-96-9
Formula:C₈H₆F₃NO₂S
MDL Number:MFCD00052206