0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
1-Bromo-2-(difluoromethyl)benzene
1-Bromo-2-(difluoromethyl)benzene
1-Bromo-2-(difluoromethyl)benzene
1-Bromo-2-(difluoromethyl)benzene
Supplier:
Apollo Scientific
Certificates
Expand image
1-Bromo-2-(difluoromethyl)benzene
Supplier:
Apollo Scientific
About this item
CAS Number:
845866-82-2
:
Formula:
C₇H₅BrF₂
MDL Number:
MFCD06657962
:
207.0154 g/mol
Boiling Pt:
62…63 °C/9mmHg
:
GHS07
Storage Temperature:
Ambient