Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Chloro-2,6-difluorobenzylamine
3-Chloro-2,6-difluorobenzylamine 98%
3-Chloro-2,6-difluorobenzylamine 98%
Corrosive
3-Chloro-2,6-difluorobenzylamine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
3-Chloro-2,6-difluorobenzylamine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
261762-46-3
:
Formula:
C₇H₆ClF₂N
MDL Number:
MFCD01631388
ADR:
8,III
:
177.5800 g/mol
UN:
2735
:
GHS05
Storage Temperature:
Ambient