Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-sec-Butyl-5-oxopyrrolidine-3-carboxylic acid
1-sec-Butyl-5-oxopyrrolidine-3-carboxylic acid 95%
1-sec-Butyl-5-oxopyrrolidine-3-carboxylic acid 95%
1-sec-Butyl-5-oxopyrrolidine-3-carboxylic acid 95%
Supplier:
Apollo Scientific
Certificates
Expand image
1-sec-Butyl-5-oxopyrrolidine-3-carboxylic acid 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
696647-92-4
:
Formula:
C₉H₁₅NO₃
MDL Number:
MFCD03834530
:
185.22 g/mol
Storage Temperature:
Ambient