tert-Butyl-5-bromo-1H-indazole-1-carboxylate
Catalog # APOSOR9627-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:297.16 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:651780-02-8
Formula:C₁₂H₁₃BrN₂O₂
MDL Number:MFCD08435913