Order Entry
Germany
ContactUsLinkComponent
N-Boc-4-carboxymethoxypiperidine 97%
N-Boc-4-carboxymethoxypiperidine 97%
Catalog # APOSOR962583-1G
Supplier:  Apollo Scientific
CAS Number:  
N-Boc-4-carboxymethoxypiperidine 97%
Catalog # APOSOR962583-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :259.30 g/mol
Storage Temperature:Ambient
CAS Number:161948-70-5
Formula:C₁₂H₂₁NO₅
MDL Number:MFCD02178987