(R)-3-(Boc-amino)-3-phenylpropionic acid 97%
Catalog # APOSOR962337-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:265.31 g/mol
Storage Temperature:Ambient
CAS Number:161024-80-2
Formula:C₁₄H₁₉NO₄
MDL Number:MFCD01320859