Order Entry
Germany
ContactUsLinkComponent
3-(tert-Butyl)-5-methyl-1H-pyrazole 97%
3-(tert-Butyl)-5-methyl-1H-pyrazole 97%
Catalog # APOSOR960601-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(tert-Butyl)-5-methyl-1H-pyrazole 97%
Catalog # APOSOR960601-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :138.21 g/mol
Storage Temperature:Ambient
CAS Number:96440-80-1
Formula:C₈H₁₄N₂
MDL Number:MFCD18325158