Order Entry
Germany
ContactUsLinkComponent
2-Amino-5-thiazolecarboxylic acid 95%
Irritant
2-Amino-5-thiazolecarboxylic acid 95%
Catalog # APOSOR9398-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Amino-5-thiazolecarboxylic acid 95%
Catalog # APOSOR9398-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :144.1500 g/mol
Melting Pt:182…184 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:40283-46-3
Formula:C₄H₄N₂O₂S
MDL Number:MFCD06203554