Acetyl iodide 95%
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:507-02-8
- Formula:C₂H₃IO
- MDL Number:MFCD00039399
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
- CAS Number:507-02-8
- Formula:C₂H₃IO
- MDL Number:MFCD00039399
- Storage Temperature:Ambient
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



