Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-(1,3-Dioxolan-2-yl)-2-furaldehyde
5-(1,3-Dioxolan-2-yl)-2-furaldehyde 95%
5-(1,3-Dioxolan-2-yl)-2-furaldehyde 95%
5-(1,3-Dioxolan-2-yl)-2-furaldehyde 95%
Supplier:
Apollo Scientific
Certificates
Expand image
5-(1,3-Dioxolan-2-yl)-2-furaldehyde 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
117953-13-6
:
Formula:
C₈H₈O₄
MDL Number:
MFCD00671521
:
168.15 g/mol
Storage Temperature:
Ambient