Order Entry
Germany
ContactUsLinkComponent
2-tert-Butyl-6-methylaniline 95%
2-tert-Butyl-6-methylaniline 95%
Catalog # APOSOR937298-1G
Supplier:  Apollo Scientific
CAS Number:  
2-tert-Butyl-6-methylaniline 95%
Catalog # APOSOR937298-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :163.26 g/mol
Storage Temperature:Ambient
CAS Number:13117-94-7
Formula:C₁₁H₁₇N
MDL Number:MFCD00025208