Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tert-Butyl methyl iminodicarboxylate
tert-Butyl methyl iminodicarboxylate 95%
tert-Butyl methyl iminodicarboxylate 95%
tert-Butyl methyl iminodicarboxylate 95%
Supplier:
Apollo Scientific
Certificates
Expand image
tert-Butyl methyl iminodicarboxylate 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
66389-76-2
:
Formula:
C₇H₁₃NO₄
MDL Number:
MFCD01321176
:
175.18 g/mol
Storage Temperature:
Ambient