Order Entry
Germany
ContactUsLinkComponent
4-(Trimethylsilyl)-3-butyn-1-ol 95%
Flammable
Irritant
4-(Trimethylsilyl)-3-butyn-1-ol 95%
Catalog # APOSOR936873-25G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Irritant
4-(Trimethylsilyl)-3-butyn-1-ol 95%
Catalog # APOSOR936873-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :142.27 g/mol
Boiling Pt:97…98 °C
Density:0.854 g/cm³ (25 °C)
Flash Pt:60 °C
UN:1993
ADR:3,III
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:2117-12-6
Formula:C₇H₁₄OSi
MDL Number:MFCD00190217