Order Entry
Germany
ContactUsLinkComponent
1,3-Diethyl-2-thiourea 95%
1,3-Diethyl-2-thiourea 95%
Catalog # APOSOR936470-100G
Supplier:  Apollo Scientific
CAS Number:  
1,3-Diethyl-2-thiourea 95%
Catalog # APOSOR936470-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :132.23 g/mol
Storage Temperature:Ambient
CAS Number:105-55-5
Formula:C₅H₁₂N₂S
MDL Number:MFCD00004925