Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Benzalbutyramide
3-Benzalbutyramide 95%
3-Benzalbutyramide 95%
3-Benzalbutyramide 95%
APOSOR936464-1G
:
Apollo Scientific
:
3-Benzalbutyramide 95%
APOSOR936464-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
175.23 g/mol
:
Ambient
:
7236-47-7
:
C₁₁H₁₃NO
:
MFCD00059860