Order Entry
Germany
ContactUsLinkComponent
Isoquinoline-3-carbonitrile 95%
Isoquinoline-3-carbonitrile 95%
Catalog # APOSOR930950-5G
Supplier:  Apollo Scientific
CAS Number:  
Isoquinoline-3-carbonitrile 95%
Catalog # APOSOR930950-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :154.17 g/mol
Storage Temperature:Ambient
CAS Number:26947-41-1
Formula:C₁₀H₆N₂
MDL Number:MFCD00075136