Butylamine hydrochloride 95%
Catalog # APOSOR930840-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:109.60 g/mol
Storage Temperature:Ambient
CAS Number:3858-78-4
Formula:C₄H₁₁N
MDL Number:MFCD00043791