Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Amino-2-bromo-5-methoxypyridine
3-Amino-2-bromo-5-methoxypyridine 98%
3-Amino-2-bromo-5-methoxypyridine 98%
Irritant
3-Amino-2-bromo-5-methoxypyridine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-Amino-2-bromo-5-methoxypyridine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
1043688-99-8
:
Formula:
C₆H₇BrN₂O
MDL Number:
MFCD16659663
:
203.0366 g/mol
:
GHS07
Storage Temperature:
Refrigerator