Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1H-Pyrrolo[2,3-b]pyridine-2-carboxaldehyde
1H-Pyrrolo[2,3-b]pyridine-2-carboxaldehyde 95%
1H-Pyrrolo[2,3-b]pyridine-2-carboxaldehyde 95%
1H-Pyrrolo[2,3-b]pyridine-2-carboxaldehyde 95%
Supplier:
Apollo Scientific
Certificates
Expand image
1H-Pyrrolo[2,3-b]pyridine-2-carboxaldehyde 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
394223-03-1
:
Formula:
C₈H₆N₂O
MDL Number:
MFCD08437634
:
146.15 g/mol
Storage Temperature:
Ambient