Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-sec-Butylaniline
2-sec-Butylaniline 98%
2-sec-Butylaniline 98%
2-sec-Butylaniline 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2-sec-Butylaniline 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
55751-54-7
:
Formula:
C₁₀H₁₅N
:
149.24 g/mol
Storage Temperature:
Ambient